Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
50 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616690 MIM: 601883 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP104 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEP104 - centrosomal protein 104 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DFFB - DNA fragmentation factor subunit beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282669.1 | Intron | NP_001269598.1 | ||||
NM_001320132.1 | Intron | NP_001307061.1 | ||||
NM_001320136.1 | Intron | NP_001307065.1 | ||||
NM_004402.3 | Intron | NP_004393.1 | ||||
XM_011540865.2 | Intron | XP_011539167.1 | ||||
XM_017000498.1 | Intron | XP_016855987.1 | ||||
XM_017000499.1 | Intron | XP_016855988.1 | ||||
XM_017000500.1 | Intron | XP_016855989.1 |