Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTACCCTCCTCCGGCGCGGCCCCT[C/T]ATCCCGGCGAGCACGGCGGCGGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605003 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SENP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SENP6 - SUMO1/sentrin specific peptidase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100409.2 | 596 | UTR 5 | NP_001093879.1 | |||
NM_001304792.1 | 596 | UTR 5 | NP_001291721.1 | |||
NM_015571.3 | 596 | UTR 5 | NP_056386.2 | |||
XM_006715457.3 | 596 | UTR 5 | XP_006715520.1 | |||
XM_011535746.2 | 596 | UTR 5 | XP_011534048.1 | |||
XM_011535747.2 | 596 | UTR 5 | XP_011534049.1 | |||
XM_017010719.1 | 596 | UTR 5 | XP_016866208.1 | |||
XM_017010720.1 | 596 | Intron | XP_016866209.1 | |||
XM_017010721.1 | 596 | Intron | XP_016866210.1 | |||
XM_017010722.1 | 596 | UTR 5 | XP_016866211.1 | |||
XM_017010723.1 | 596 | UTR 5 | XP_016866212.1 | |||
XM_017010724.1 | 596 | UTR 5 | XP_016866213.1 | |||
XM_017010725.1 | 596 | UTR 5 | XP_016866214.1 |