Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCATGTGTTCACAAGCTTCTAAAA[C/T]GTTTGCCAAAATTAAAGTCTGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606489 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXOSC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EXOSC3 - exosome component 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002269.2 | 805 | UTR 3 | NP_001002269.1 | |||
NM_016042.3 | 805 | Missense Mutation | ATT,GTT | I,V 253 | NP_057126.2 |
LOC105376037 - uncharacterized LOC105376037 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRMT10B - tRNA methyltransferase 10B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286950.1 | 805 | Intron | NP_001273879.1 | |||
NM_001286951.1 | 805 | Intron | NP_001273880.1 | |||
NM_001286952.1 | 805 | Intron | NP_001273881.1 | |||
NM_001286953.1 | 805 | Intron | NP_001273882.1 | |||
NM_001286954.1 | 805 | Intron | NP_001273883.1 | |||
NM_144964.3 | 805 | Intron | NP_659401.2 | |||
XM_005251375.2 | 805 | Intron | XP_005251432.1 | |||
XM_005251379.2 | 805 | Intron | XP_005251436.1 | |||
XM_011517735.2 | 805 | Intron | XP_011516037.1 | |||
XM_011517736.2 | 805 | Intron | XP_011516038.1 | |||
XM_011517738.2 | 805 | Intron | XP_011516040.1 | |||
XM_011517739.2 | 805 | Intron | XP_011516041.1 | |||
XM_011517742.1 | 805 | Intron | XP_011516044.1 | |||
XM_011517743.2 | 805 | Intron | XP_011516045.1 | |||
XM_017014313.1 | 805 | Intron | XP_016869802.1 |