Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTCTCCTATGTGGGAAACAATTT[C/T]GGGACTGGAGCCCTCAAATGCAACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
POLR1E PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
POLR1E - RNA polymerase I subunit E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282766.1 | 335 | UTR 5 | NP_001269695.1 | |||
NM_022490.2 | 335 | Silent Mutation | TTC,TTT | F,F 74 | NP_071935.1 | |
XM_005251547.2 | 335 | Silent Mutation | TTC,TTT | F,F 136 | XP_005251604.1 |
Set Membership: |
HapMap |