Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604148 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC13A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC13A2 - solute carrier family 13 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145975.1 | Intron | NP_001139447.1 | ||||
NM_003984.3 | Intron | NP_003975.1 | ||||
XM_006722165.2 | Intron | XP_006722228.1 | ||||
XM_011525450.2 | Intron | XP_011523752.1 | ||||
XM_011525451.1 | Intron | XP_011523753.1 | ||||
XM_011525452.1 | Intron | XP_011523754.1 | ||||
XM_011525453.2 | Intron | XP_011523755.1 | ||||
XM_011525454.2 | Intron | XP_011523756.1 |