Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACATGCACTGCACAGGTTCCGCTC[C/T]CGTCAGAAGGTGGGCTCCCCCACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611519 MIM: 616815 MIM: 191161 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4723 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR4723 - microRNA 4723 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLDIP2 - DNA polymerase delta interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290145.1 | Intron | NP_001277074.1 | ||||
NM_015584.4 | Intron | NP_056399.1 |
TMEM199 - transmembrane protein 199 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNFAIP1 - TNF alpha induced protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |