Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCATATACACTTGCCTCAAGGCCT[A/G]TGTCATCGAGGAGCCACCGGAGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608461 | ||||||||||||||||||||
Literature Links: |
COL27A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COL27A1 - collagen type XXVII alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032888.3 | Intron | NP_116277.2 | ||||
XM_006717308.2 | Intron | XP_006717371.1 | ||||
XM_006717310.3 | Intron | XP_006717373.1 | ||||
XM_011519138.2 | Intron | XP_011517440.1 | ||||
XM_011519140.1 | Intron | XP_011517442.1 | ||||
XM_011519142.2 | Intron | XP_011517444.1 | ||||
XM_011519143.2 | Intron | XP_011517445.1 | ||||
XM_011519144.2 | Intron | XP_011517446.1 | ||||
XM_011519145.2 | Intron | XP_011517447.1 | ||||
XM_017015239.1 | Intron | XP_016870728.1 |
MIR455 - microRNA 455 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |