Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATAGGTTCTCCTAGTGTCTATCA[C/T]GCAGTTATCGTCATCTTTTTGGAGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
47 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MFSD14A PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
MFSD14A - major facilitator superfamily domain containing 14A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033055.2 | 360 | Silent Mutation | CAC,CAT | H,H 39 | NP_149044.2 | |
XM_005271118.1 | 360 | Silent Mutation | CAC,CAT | H,H 17 | XP_005271175.1 | |
XM_011541957.1 | 360 | Silent Mutation | CAC,CAT | H,H 39 | XP_011540259.1 | |
XM_017002084.1 | 360 | Silent Mutation | CAC,CAT | H,H 17 | XP_016857573.1 |