Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGTCCAGGACTGGGTGGGTTGGG[C/T]GGAGCTGGACTTAGCCCCAGGCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 173310 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAEP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAEP - progestagen associated endometrial protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018048.1 | Intron | NP_001018058.1 | ||||
NM_001018049.2 | Intron | NP_001018059.1 | ||||
NM_002571.3 | Intron | NP_002562.2 | ||||
XM_011518745.2 | Intron | XP_011517047.1 | ||||
XM_011518746.2 | Intron | XP_011517048.1 | ||||
XM_011518747.2 | Intron | XP_011517049.1 | ||||
XM_011518748.1 | Intron | XP_011517050.1 | ||||
XM_011518749.2 | Intron | XP_011517051.1 | ||||
XM_011518751.1 | Intron | XP_011517053.1 | ||||
XM_011518752.2 | Intron | XP_011517054.1 | ||||
XM_017014782.1 | Intron | XP_016870271.1 | ||||
XM_017014783.1 | Intron | XP_016870272.1 |