Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATGACTTAAAGTTGCAAACCTTT[A/G]AGAAGCTTCTTCAGTAGGTCATCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611342 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf64 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C9orf64 - chromosome 9 open reading frame 64 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317997.1 | 1031 | Silent Mutation | CTC,CTT | L,L 122 | NP_001304926.1 | |
NM_032307.4 | 1031 | Silent Mutation | CTC,CTT | L,L 263 | NP_115683.3 | |
XM_005252271.4 | 1031 | Intron | XP_005252328.1 |