Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCTTGCTCACACTGACCTGGGAG[C/T]TCTGGGAGCAGGGCCTGGTCAGGCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602311 MIM: 607028 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGRP PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
|||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
AGRP - agouti related neuropeptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001138.1 | 423 | Silent Mutation | GAA,GAG | E,E 41 | NP_001129.1 |
ATP6V0D1 - ATPase H+ transporting V0 subunit d1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |