Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGCACTGATCCACTGCTGGACA[A/G]ACCCGGGGCAGCCTCTGGGTGAACA
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605278 MIM: 614778 | ||||||||||||||||||||||||||||||||
Literature Links: |
CES2 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian
|
CEPH (CEU) - Not Available | |||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | |||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CES2 - carboxylesterase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003869.5 | 1124 | Missense Mutation | AAC,GAC | N,D 42 | NP_003860.2 | |
NM_198061.2 | 1124 | Missense Mutation | AAC,GAC | N,D 42 | NP_932327.1 | |
XM_011523421.2 | 1124 | UTR 5 | XP_011521723.1 | |||
XM_017023818.1 | 1124 | UTR 5 | XP_016879307.1 |
FAM96B - family with sequence similarity 96 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
DME Validated Inventoried |