Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGATGACCAAAGACAGCCTTGCC[C/T]TTTTCCGTGCCAGCGCGGACAGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604885 MIM: 612584 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYBBP1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYBBP1A - MYB binding protein 1a | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105538.1 | 3958 | Missense Mutation | AAG,AGG | K,R 1299 | NP_001099008.1 | |
NM_014520.3 | 3958 | Missense Mutation | AAG,AGG | K,R 1299 | NP_055335.2 | |
XM_011523616.2 | 3958 | Missense Mutation | AAG,AGG | K,R 1047 | XP_011521918.1 |
SPNS2 - sphingolipid transporter 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |