Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTTTTCCTGGTGGTTGATTGGT[A/G]TATAAAAGGGAAGATAATTCAAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606193 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC13A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC13A1 - solute carrier family 13 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324400.1 | Intron | NP_001311329.1 | ||||
NM_022444.3 | Intron | NP_071889.2 | ||||
XM_011516515.2 | Intron | XP_011514817.1 | ||||
XM_011516516.2 | Intron | XP_011514818.1 | ||||
XM_011516517.2 | Intron | XP_011514819.1 | ||||
XM_011516518.2 | Intron | XP_011514820.1 | ||||
XM_011516519.2 | Intron | XP_011514821.1 | ||||
XM_017012554.1 | Intron | XP_016868043.1 | ||||
XM_017012555.1 | Intron | XP_016868044.1 |
Set Membership: |
HapMap |