Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTACTCTTTAATCTTAAAATAGC[C/T]TTGAGATACACGTAATCAAGTAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606478 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
POT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
POT1 - protection of telomeres 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042594.1 | Intron | NP_001036059.1 | ||||
NM_015450.2 | Intron | NP_056265.2 | ||||
XM_006715917.3 | Intron | XP_006715980.1 | ||||
XM_011516006.1 | Intron | XP_011514308.1 | ||||
XM_011516007.1 | Intron | XP_011514309.1 | ||||
XM_017011941.1 | Intron | XP_016867430.1 | ||||
XM_017011942.1 | Intron | XP_016867431.1 |