Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTTAGTTTTCTTTACAAATGAAA[A/C]CATAAAATATAAAGGATATAAGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 168470 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTHLH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTHLH - parathyroid hormone like hormone | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002820.2 | Intron | NP_002811.1 | ||||
NM_198964.1 | Intron | NP_945315.1 | ||||
NM_198965.1 | Intron | NP_945316.1 | ||||
NM_198966.1 | Intron | NP_945317.1 | ||||
XM_011520774.2 | Intron | XP_011519076.1 | ||||
XM_011520775.2 | Intron | XP_011519077.1 | ||||
XM_017019674.1 | Intron | XP_016875163.1 | ||||
XM_017019675.1 | Intron | XP_016875164.1 |