Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAATATCTGTATTAAGGCTGAGG[C/T]AAAGAGAAAAGACACGGCTATATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603501 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PARG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PARG - poly(ADP-ribose) glycohydrolase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303486.1 | Intron | NP_001290415.1 | ||||
NM_001303487.1 | Intron | NP_001290416.1 | ||||
NM_001303489.1 | Intron | NP_001290418.1 | ||||
NM_001324381.1 | Intron | NP_001311310.1 | ||||
NM_003631.3 | Intron | NP_003622.2 | ||||
XM_011540305.1 | Intron | XP_011538607.1 | ||||
XM_011540308.2 | Intron | XP_011538610.1 |