Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTCAACAGGCATGCTCCAACTGC[A/C]CCGAAGTCAAGAGCACTCCTTCTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608533 MIM: 602164 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO38 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXO38 - F-box protein 38 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HTR4 - 5-hydroxytryptamine receptor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000870.6 | Intron | NP_000861.1 | ||||
NM_001040169.2 | Intron | NP_001035259.1 | ||||
NM_001040172.2 | Intron | NP_001035262.2 | ||||
NM_001040173.2 | Intron | NP_001035263.1 | ||||
NM_001286410.1 | Intron | NP_001273339.1 | ||||
NM_199453.3 | Intron | NP_955525.1 |