Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTCCCTTAAGAATTTTCCCAAAC[C/T]GGCAGCACCAAGGTAGATGAAAACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 153615 MIM: 615427 | ||||||||||||||||||||
Literature Links: |
CAPG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAPG - capping actin protein, gelsolin like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256139.1 | Intron | NP_001243068.1 | ||||
NM_001256140.1 | Intron | NP_001243069.1 | ||||
NM_001320732.1 | Intron | NP_001307661.1 | ||||
NM_001320733.1 | Intron | NP_001307662.1 | ||||
NM_001320734.1 | Intron | NP_001307663.1 | ||||
NM_001747.3 | Intron | NP_001738.2 |
ELMOD3 - ELMO domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |