Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATTATGGATACTAACATGTTAA[C/T]GTTAGTATAGCATATTGTTTTTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602731 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FYB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FYB - FYN binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243093.1 | Intron | NP_001230022.1 | ||||
NM_001465.4 | Intron | NP_001456.3 | ||||
NM_199335.3 | Intron | NP_955367.1 | ||||
XM_006714464.2 | Intron | XP_006714527.1 | ||||
XM_006714465.2 | Intron | XP_006714528.1 | ||||
XM_006714466.2 | Intron | XP_006714529.1 | ||||
XM_011514008.2 | Intron | XP_011512310.1 | ||||
XM_011514009.1 | Intron | XP_011512311.1 | ||||
XM_011514010.1 | Intron | XP_011512312.1 | ||||
XM_011514011.2 | Intron | XP_011512313.1 | ||||
XM_011514012.2 | Intron | XP_011512314.1 | ||||
XM_011514013.2 | Intron | XP_011512315.1 | ||||
XM_017009316.1 | Intron | XP_016864805.1 |