Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCTGGTGGGTGGGGCCCTGGCA[C/G]GCAATCGTCCGCATCTTCCACCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601270 | ||||||||||||||||||||||||||||||||
Literature Links: |
CYP4F3 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian
|
CEPH (CEU) - Not Available | |||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | |||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CYP4F3 - cytochrome P450 family 4 subfamily F member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000896.2 | 90 | Silent Mutation | CAC,CAG | H,Q 96 | NP_000887.2 | |
NM_001199208.1 | 90 | Intron | NP_001186137.1 | |||
NM_001199209.1 | 90 | Intron | NP_001186138.1 | |||
XM_005259911.4 | 90 | Intron | XP_005259968.1 | |||
XM_011528014.2 | 90 | UTR 5 | XP_011526316.1 | |||
XM_017026814.1 | 90 | Intron | XP_016882303.1 | |||
XM_017026815.1 | 90 | Intron | XP_016882304.1 |
Set Membership: |
DME Validated Inventoried |