Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGCTGCTGGCCTGTCACTGGTC[A/G]AGGTGGTCCGGGGATTCACGCCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602171 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPA33 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPA33 - glycoprotein A33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005814.2 | 1123 | Silent Mutation | CTC,CTT | L,L 317 | NP_005805.1 | |
XM_017000005.1 | 1123 | Silent Mutation | CTC,CTT | L,L 209 | XP_016855494.1 |