Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
43 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600275 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOTCH2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NOTCH2 - notch 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001200001.1 | 7638 | Intron | NP_001186930.1 | |||
NM_024408.3 | 7638 | Silent Mutation | GGA,GGT | G,G 2447 | NP_077719.2 | |
XM_005270901.3 | 7638 | Silent Mutation | GGA,GGT | G,G 2408 | XP_005270958.1 | |
XM_011541519.2 | 7638 | Silent Mutation | GGA,GGT | G,G 2443 | XP_011539821.1 | |
XM_011541520.2 | 7638 | Silent Mutation | GGA,GGT | G,G 2408 | XP_011539822.1 | |
XM_017001372.1 | 7638 | Silent Mutation | GGA,GGT | G,G 2431 | XP_016856861.1 | |
XM_017001373.1 | 7638 | Silent Mutation | GGA,GGT | G,G 2408 | XP_016856862.1 |