Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGGTCAGGCCTTGTAGGGGGCTT[A/G]GCACTAAAGGCAGAGTTCTGAACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609885 MIM: 602046 MIM: 605054 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ELL3 - elongation factor for RNA polymerase II 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025165.2 | 1116 | Missense Mutation | CCA,CTA | P,L 238 | NP_079441.1 |
PDIA3 - protein disulfide isomerase family A member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SERF2 - small EDRK-rich factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |