Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613110 MIM: 603106 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BLCAP PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BLCAP - bladder cancer associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167820.1 | 900 | Intron | NP_001161292.1 | |||
NM_001167821.1 | 900 | Intron | NP_001161293.1 | |||
NM_001167822.2 | 900 | Intron | NP_001161294.1 | |||
NM_001167823.1 | 900 | Intron | NP_001161295.1 | |||
NM_001317074.1 | 900 | Intron | NP_001304003.1 | |||
NM_001317075.1 | 900 | Intron | NP_001304004.1 | |||
NM_006698.3 | 900 | Intron | NP_006689.1 | |||
XM_017027599.1 | 900 | UTR 5 | XP_016883088.1 |
NNAT - neuronatin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322802.1 | 900 | Intron | NP_001309731.1 | |||
NM_005386.3 | 900 | Intron | NP_005377.1 | |||
NM_181689.2 | 900 | Intron | NP_859017.1 |