Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAGATTGAGACAAACCATTGTGT[A/G]GTTGGGTTTGGGTCAGCAGGCTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605096 MIM: 142695 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANO7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANO7 - anoctamin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001666.3 | 4084 | Intron | NP_001001666.1 | |||
NM_001001891.3 | 4084 | Intron | NP_001001891.2 | |||
XM_011511263.2 | 4084 | Intron | XP_011509565.1 | |||
XM_011511267.2 | 4084 | Intron | XP_011509569.1 | |||
XM_017004229.1 | 4084 | Intron | XP_016859718.1 | |||
XM_017004230.1 | 4084 | Intron | XP_016859719.1 |
HDLBP - high density lipoprotein binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243900.2 | 4084 | UTR 3 | NP_001230829.1 | |||
NM_001320965.1 | 4084 | UTR 3 | NP_001307894.1 | |||
NM_001320966.1 | 4084 | UTR 3 | NP_001307895.1 | |||
NM_001320967.1 | 4084 | Intron | NP_001307896.1 | |||
NM_005336.5 | 4084 | UTR 3 | NP_005327.1 | |||
NM_203346.4 | 4084 | UTR 3 | NP_976221.1 | |||
XM_005247002.3 | 4084 | UTR 3 | XP_005247059.2 | |||
XM_005247003.4 | 4084 | UTR 3 | XP_005247060.2 | |||
XM_006712475.3 | 4084 | UTR 3 | XP_006712538.1 | |||
XM_011511058.2 | 4084 | UTR 3 | XP_011509360.1 | |||
XM_011511060.2 | 4084 | UTR 3 | XP_011509362.1 | |||
XM_017003940.1 | 4084 | UTR 3 | XP_016859429.1 | |||
XM_017003941.1 | 4084 | Intron | XP_016859430.1 | |||
XM_017003942.1 | 4084 | Intron | XP_016859431.1 |