Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611386 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADNP PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ADNP - activity dependent neuroprotector homeobox | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282531.1 | 4594 | UTR 3 | NP_001269460.1 | |||
NM_001282532.1 | 4594 | UTR 3 | NP_001269461.1 | |||
NM_015339.3 | 4594 | UTR 3 | NP_056154.1 | |||
NM_181442.2 | 4594 | UTR 3 | NP_852107.1 | |||
XM_011528747.2 | 4594 | UTR 3 | XP_011527049.1 | |||
XM_011528748.2 | 4594 | UTR 3 | XP_011527050.1 | |||
XM_017027757.1 | 4594 | UTR 3 | XP_016883246.1 | |||
XM_017027758.1 | 4594 | UTR 3 | XP_016883247.1 | |||
XM_017027759.1 | 4594 | UTR 3 | XP_016883248.1 |