Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCAGCAGGCCAGTTTATCTCCCT[A/G]CTTTCCTGTCCACATAGGCAATAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615762 MIM: 613377 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRXCR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRXCR2 - glutaredoxin and cysteine rich domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH3RF2 - SH3 domain containing ring finger 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152550.3 | Intron | NP_689763.3 | ||||
XM_005268380.2 | Intron | XP_005268437.1 | ||||
XM_006714757.1 | Intron | XP_006714820.1 | ||||
XM_011537567.2 | Intron | XP_011535869.1 | ||||
XM_011537568.2 | Intron | XP_011535870.1 | ||||
XM_011537569.2 | Intron | XP_011535871.1 | ||||
XM_017009136.1 | Intron | XP_016864625.1 |