Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601828 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NR4A2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
NR4A2 - nuclear receptor subfamily 4 group A member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006186.3 | Intron | NP_006177.1 | ||||
XM_005246621.3 | Intron | XP_005246678.1 | ||||
XM_005246622.3 | Intron | XP_005246679.1 | ||||
XM_006712553.3 | Intron | XP_006712616.1 | ||||
XM_011511246.1 | Intron | XP_011509548.1 | ||||
XM_017004219.1 | Intron | XP_016859708.1 | ||||
XM_017004220.1 | Intron | XP_016859709.1 |
Set Membership: |
HapMap |