Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCACTGGAGTGGCAGATAGTTGCC[A/G]TTCTCGTCGCACTTGGGCCTGAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142790 MIM: 606847 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD74 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD74 - CD74 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025158.2 | 868 | Intron | NP_001020329.1 | |||
NM_001025159.2 | 868 | Silent Mutation | AAC,AAT | N,N 235 | NP_001020330.1 | |
NM_004355.3 | 868 | Intron | NP_004346.1 | |||
XM_017010089.1 | 868 | Silent Mutation | AAC,AAT | N,N 235 | XP_016865578.1 | |
XM_017010090.1 | 868 | Intron | XP_016865579.1 |
TCOF1 - treacle ribosome biogenesis factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |