Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGTGCCCCATGGGAGTTCTATTG[C/T]GGTAGGGCTTAATGAAGATGACTCG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612979 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYS1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SYS1 - Sys1 golgi trafficking protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099791.2 | 912 | UTR 3 | NP_001093261.1 | |||
NM_001197129.1 | 912 | Intron | NP_001184058.1 | |||
NM_033542.3 | 912 | Intron | NP_291020.1 |
SYS1-DBNDD2 - SYS1-DBNDD2 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TP53TG5 - TP53 target 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014477.2 | 912 | Missense Mutation | CAC,CGC | H,R 191 | NP_055292.1 | |
XM_011528790.2 | 912 | Missense Mutation | CAC,CGC | H,R 175 | XP_011527092.1 |
Set Membership: |
HapMap |