Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCGCAGCCATGGCGGCCGCCGCG[A/C]TCCCAGCATGGCTGTCTCTGCAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611995 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPS35 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRPS35 - mitochondrial ribosomal protein S35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190864.1 | 87 | Missense Mutation | ATC,CTC | I,L 6 | NP_001177793.1 | |
NM_021821.3 | 87 | Missense Mutation | ATC,CTC | I,L 6 | NP_068593.2 | |
XM_017019780.1 | 87 | Missense Mutation | ATC,CTC | I,L 6 | XP_016875269.1 |