Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATCTTCTTTGCTTTAATTGGTGT[G/T]ATGTCTCCATTTTCATCTTCTTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605500 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPHOSPH6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MPHOSPH6 - M-phase phosphoprotein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005792.2 | 601 | Silent Mutation | ATA,ATC | I,I 146 | NP_005783.2 | |
XM_011522808.2 | 601 | Silent Mutation | ATA,ATC | I,I 128 | XP_011521110.1 | |
XM_017022852.1 | 601 | Silent Mutation | ATA,ATC | I,I 128 | XP_016878341.1 |