Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGAGCTGGTAGCTGCGAGGTCA[C/T]GCTCAGCCTGTGTACGTGGTGCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611411 MIM: 600845 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAMKK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAMKK1 - calcium/calmodulin dependent protein kinase kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
P2RX1 - purinergic receptor P2X 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002558.3 | 1583 | Missense Mutation | CAT,CGT | H,R 381 | NP_002549.1 | |
XM_006721529.2 | 1583 | Missense Mutation | CAT,CGT | H,R 364 | XP_006721592.1 | |
XM_011523896.2 | 1583 | Missense Mutation | CAT,CGT | H,R 413 | XP_011522198.1 | |
XM_011523897.2 | 1583 | Missense Mutation | CAT,CGT | H,R 401 | XP_011522199.1 | |
XM_011523898.2 | 1583 | Missense Mutation | CAT,CGT | H,R 396 | XP_011522200.1 | |
XM_011523899.2 | 1583 | Missense Mutation | CAT,CGT | H,R 393 | XP_011522201.1 | |
XM_011523900.2 | 1583 | Intron | XP_011522202.1 |