Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 190196 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TGM2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TGM2 - transglutaminase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323316.1 | 2137 | Silent Mutation | NP_001310245.1 | |||
NM_001323317.1 | 2137 | Silent Mutation | NP_001310246.1 | |||
NM_001323318.1 | 2137 | Silent Mutation | NP_001310247.1 | |||
NM_004613.3 | 2137 | Silent Mutation | NP_004604.2 | |||
NM_198951.2 | 2137 | Intron | NP_945189.1 | |||
XM_011529028.1 | 2137 | Silent Mutation | XP_011527330.1 |
Set Membership: |
HapMap |