Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAAGTTAAAGCGCCCTCTGCCAC[A/G]TTGAAAAGTACCACGACCTCTTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612588 | ||||||||||||||||||||
Literature Links: |
BCLAF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BCLAF1 - BCL2 associated transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077440.1 | 3027 | Missense Mutation | CGT,TGT | R,C 820 | NP_001070908.1 | |
NM_001077441.1 | 3027 | Missense Mutation | CGT,TGT | R,C 698 | NP_001070909.1 | |
NM_001301038.1 | 3027 | Missense Mutation | CGT,TGT | R,C 869 | NP_001287967.1 | |
NM_014739.2 | 3027 | Missense Mutation | CGT,TGT | R,C 871 | NP_055554.1 | |
XM_005267237.3 | 3027 | Missense Mutation | CGT,TGT | R,C 822 | XP_005267294.1 | |
XM_017011529.1 | 3027 | Missense Mutation | CGT,TGT | R,C 696 | XP_016867018.1 | |
XM_017011530.1 | 3027 | Missense Mutation | CGT,TGT | R,C 649 | XP_016867019.1 | |
XM_017011531.1 | 3027 | Missense Mutation | CGT,TGT | R,C 647 | XP_016867020.1 | |
XM_017011532.1 | 3027 | Missense Mutation | CGT,TGT | R,C 260 | XP_016867021.1 |
MTFR2 - mitochondrial fission regulator 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |