Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCCAGGATGAGGATCTGGGTCTT[T/C]CGGAGAAGGGCACGTGCCAGACACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 158343 MIM: 603234 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCC1 - ATP binding cassette subfamily C member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ABCC6 - ATP binding cassette subfamily C member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079528.3 | 4267 | Intron | NP_001072996.1 | |||
NM_001171.5 | 4267 | Silent Mutation | CGA,CGG | R,R 1418 | NP_001162.4 | |
XM_011522479.2 | 4267 | Silent Mutation | CGA,CGG | R,R 1407 | XP_011520781.1 | |
XM_011522480.1 | 4267 | Silent Mutation | CGA,CGG | R,R 1304 | XP_011520782.1 | |
XM_011522481.2 | 4267 | Silent Mutation | CGA,CGG | R,R 1304 | XP_011520783.1 | |
XM_011522482.2 | 4267 | Intron | XP_011520784.1 | |||
XM_017023212.1 | 4267 | Silent Mutation | CGA,CGG | R,R 1362 | XP_016878701.1 | |
XM_017023213.1 | 4267 | Silent Mutation | CGA,CGG | R,R 1304 | XP_016878702.1 | |
XM_017023214.1 | 4267 | Intron | XP_016878703.1 |