Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 126337 MIM: 156560 MIM: 614489 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDIT3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DDIT3 - DNA damage inducible transcript 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195053.1 | 601 | Silent Mutation | AGC,AGT | S,S 111 | NP_001181982.1 | |
NM_001195054.1 | 601 | Silent Mutation | AGC,AGT | S,S 111 | NP_001181983.1 | |
NM_001195055.1 | 601 | Silent Mutation | AGC,AGT | S,S 111 | NP_001181984.1 | |
NM_001195056.1 | 601 | Silent Mutation | AGC,AGT | S,S 111 | NP_001181985.1 | |
NM_001195057.1 | 601 | Silent Mutation | AGC,AGT | S,S 88 | NP_001181986.1 | |
NM_004083.5 | 601 | Silent Mutation | AGC,AGT | S,S 88 | NP_004074.2 |
MARS - methionyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004990.3 | 601 | Intron | NP_004981.2 | |||
XM_006719398.3 | 601 | Intron | XP_006719461.1 |
MBD6 - methyl-CpG binding domain protein 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR616 - microRNA 616 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6758 - microRNA 6758 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |