Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCATACCTTCTGGAGGGACGTC[A/G]ATGGTATTAGGATAGAAGCACCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 113710 MIM: 605511 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TFF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TFF1 - trefoil factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003225.2 | 262 | Silent Mutation | ATC,ATT | I,I 74 | NP_003216.1 |
TMPRSS3 - transmembrane protease, serine 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |