Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609677 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OS9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
OS9 - OS9, endoplasmic reticulum lectin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017956.2 | Intron | NP_001017956.1 | ||||
NM_001017957.2 | Intron | NP_001017957.1 | ||||
NM_001017958.2 | Intron | NP_001017958.1 | ||||
NM_001261420.1 | Intron | NP_001248349.1 | ||||
NM_001261421.1 | Intron | NP_001248350.1 | ||||
NM_001261422.1 | Intron | NP_001248351.1 | ||||
NM_001261423.1 | Intron | NP_001248352.1 | ||||
NM_006812.3 | Intron | NP_006803.1 | ||||
XM_005268581.1 | Intron | XP_005268638.1 | ||||
XM_006719200.1 | Intron | XP_006719263.1 | ||||
XM_006719201.1 | Intron | XP_006719264.1 |