Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAATCCCCAACGGCGCATTTCCAA[A/C]CCCCAGGCCTGACCTGCAGGTTCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
31 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611889 MIM: 603623 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
S100PBP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
S100PBP - S100P binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256121.1 | Intron | NP_001243050.1 | ||||
NM_022753.3 | Intron | NP_073590.2 | ||||
XM_011541961.2 | Intron | XP_011540263.1 | ||||
XM_011541962.2 | Intron | XP_011540264.1 | ||||
XM_011541963.1 | Intron | XP_011540265.1 | ||||
XM_017002098.1 | Intron | XP_016857587.1 |
YARS - tyrosyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003680.3 | Intron | NP_003671.1 | ||||
XM_011542347.1 | Intron | XP_011540649.1 | ||||
XM_011542348.1 | Intron | XP_011540650.1 | ||||
XM_017002651.1 | Intron | XP_016858140.1 |