Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTTCCCAGCCCTCAGCAACACC[A/G]GCAAGCTCGTAGTGCTTTTTCCGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601767 | ||||||||||||||||||||
Literature Links: |
HIP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HIP1 - huntingtin interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243198.2 | 3094 | Silent Mutation | GCC,GCT | A,A 961 | NP_001230127.1 | |
NM_005338.6 | 3094 | Silent Mutation | GCC,GCT | A,A 1012 | NP_005329.3 | |
XM_005250304.2 | 3094 | Silent Mutation | GCC,GCT | A,A 983 | XP_005250361.1 | |
XM_005250305.2 | 3094 | Silent Mutation | GCC,GCT | A,A 978 | XP_005250362.1 | |
XM_011516116.2 | 3094 | Silent Mutation | GCC,GCT | A,A 1012 | XP_011514418.1 | |
XM_017012099.1 | 3094 | Silent Mutation | GCC,GCT | A,A 998 | XP_016867588.1 |