Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCTCGGCGGCCCAGGGCAGCACC[G/C]AAGGTCTGGGCAAACTCAGCATTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616162 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KHNYN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KHNYN - KH and NYN domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290256.1 | 688 | UTR 3 | NP_001277185.1 | |||
NM_001290257.1 | 688 | UTR 3 | NP_001277186.1 | |||
NM_015299.2 | 688 | UTR 3 | NP_056114.1 | |||
XM_005267474.4 | 688 | UTR 3 | XP_005267531.1 | |||
XM_011536590.2 | 688 | UTR 3 | XP_011534892.1 | |||
XM_017021114.1 | 688 | Intron | XP_016876603.1 |
LOC101927045 - uncharacterized LOC101927045 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SDR39U1 - short chain dehydrogenase/reductase family 39U member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290292.1 | 688 | Missense Mutation | TTC,TTG | F,L 150 | NP_001277221.1 | |
NM_001290293.2 | 688 | Missense Mutation | TTC,TTG | F,L 124 | NP_001277222.1 | |
NM_001308075.1 | 688 | Missense Mutation | TTC,TTG | F,L 150 | NP_001295004.1 | |
NM_020195.2 | 688 | Missense Mutation | TTC,TTG | F,L 232 | NP_064580.2 | |
XM_011536975.1 | 688 | Missense Mutation | TTC,TTG | F,L 255 | XP_011535277.1 | |
XM_011536976.1 | 688 | Missense Mutation | TTC,TTG | F,L 190 | XP_011535278.1 |
Set Membership: |
HapMap |