Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASS4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CASS4 - Cas scaffolding protein family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164114.1 | 2818 | Silent Mutation | AAC,AAT | N,N 677 | NP_001157586.1 | |
NM_001164115.1 | 2818 | Silent Mutation | AAC,AAT | N,N 294 | NP_001157587.1 | |
NM_001164116.1 | 2818 | Silent Mutation | AAC,AAT | N,N 731 | NP_001157588.1 | |
NM_020356.3 | 2818 | Silent Mutation | AAC,AAT | N,N 731 | NP_065089.2 | |
XM_006723831.3 | 2818 | Silent Mutation | AAC,AAT | N,N 677 | XP_006723894.1 |