Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCTCCAGAGGCCCCCACAAAACC[A/G]GGAAAGGAAGGAAAGGACAGCGGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603369 MIM: 604460 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDKN2C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDKN2C - cyclin dependent kinase inhibitor 2C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001262.2 | 558 | UTR 5 | NP_001253.1 | |||
NM_078626.2 | 558 | Intron | NP_523240.1 |
FAF1 - Fas associated factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |