Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCTTACTCCACCCCCGTTTTGTT[A/C]CAAAGGCAAGTTCAGTGGTCTCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608335 MIM: 610035 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105371433 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105371433 - uncharacterized LOC105371433 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLEKHO1 - pleckstrin homology domain containing O1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304722.1 | Intron | NP_001291651.1 | ||||
NM_001304723.1 | Intron | NP_001291652.1 | ||||
NM_001304724.1 | Intron | NP_001291653.1 | ||||
NM_016274.5 | Intron | NP_057358.2 | ||||
XM_017001420.1 | Intron | XP_016856909.1 |
VPS45 - vacuolar protein sorting 45 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |