Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGCAAGTGCTGGACGGTAACTAC[C/G]TTTTTTGTTATTATGATGGGCTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607568 MIM: 251170 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MMAB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MMAB - methylmalonic aciduria (cobalamin deficiency) cblB type | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MVK - mevalonate kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000431.3 | Intron | NP_000422.1 | ||||
NM_001114185.2 | Intron | NP_001107657.1 | ||||
NM_001301182.1 | Intron | NP_001288111.1 | ||||
XM_011538372.2 | Intron | XP_011536674.2 | ||||
XM_017019313.1 | Intron | XP_016874802.1 | ||||
XM_017019314.1 | Intron | XP_016874803.1 |