Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601620 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBX5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TBX5 - T-box 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000192.3 | 3699 | Silent Mutation | TCC,TCT | S,S 427 | NP_000183.2 | |
NM_080717.2 | 3699 | Silent Mutation | TCC,TCT | S,S 377 | NP_542448.1 | |
NM_181486.2 | 3699 | Silent Mutation | TCC,TCT | S,S 427 | NP_852259.1 | |
XM_017019912.1 | 3699 | Silent Mutation | TCC,TCT | S,S 443 | XP_016875401.1 |