Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCATTCACAGACACCCCCATCCCT[A/G]CCCACAGCCTGCCCCCTCAGCATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
25 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606644 MIM: 600932 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGSF8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGSF8 - immunoglobulin superfamily member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206665.2 | Intron | NP_001193594.1 | ||||
NM_001320247.1 | Intron | NP_001307176.1 | ||||
NM_052868.5 | Intron | NP_443100.1 | ||||
XM_006711631.3 | Intron | XP_006711694.1 | ||||
XM_011510159.2 | Intron | XP_011508461.1 | ||||
XM_017002835.1 | Intron | XP_016858324.1 | ||||
XM_017002836.1 | Intron | XP_016858325.1 | ||||
XM_017002837.1 | Intron | XP_016858326.1 | ||||
XM_017002838.1 | Intron | XP_016858327.1 | ||||
XM_017002839.1 | Intron | XP_016858328.1 | ||||
XM_017002840.1 | Intron | XP_016858329.1 |
KCNJ9 - potassium voltage-gated channel subfamily J member 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |