Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607784 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCG4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCG4 - ATP binding cassette subfamily G member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142505.1 | Intron | NP_001135977.1 | ||||
NM_022169.4 | Intron | NP_071452.2 | ||||
XM_011542952.2 | Intron | XP_011541254.1 | ||||
XM_017018121.1 | Intron | XP_016873610.1 |